Assay Details
| Gene Symbol: SS18L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145186.2 | NP_001138658.1 | 1 | 119 | 83 | |
| GenBank mRNA | FJ546082.1 | - | 1 | 120 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SS18L2| Gene Symbol: SS18L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145186.2 | NP_001138658.1 | 1 | 119 | 83 | |
| GenBank mRNA | FJ546082.1 | - | 1 | 120 | 83 | |
Set Membership: |
|