Assay Details
| Gene Symbol: PRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163050.1 | NP_001156522.1 | 1 - 2 | 111 | 99 | |
| GenBank mRNA | FJ900271.1 | - | 1 - 2 | 111 | 99 | |
More Information
Set Membership: |
|

PRM1
Genomic Map