Assay Details
| Gene Symbol: SCD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112815.1 | NP_001106286.1 | 3 - 4 | 829 | 74 | |
| GenBank mRNA | EU272818.1 | - | 3 - 4 | 829 | 74 | |
More Information
Set Membership: |
|

SCD5
Genomic Map