Assay Details
| Gene Symbol: SCD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112815.1 | NP_001106286.1 | 4 - 5 | 1066 | 79 | |
| GenBank mRNA | EU272818.1 | - | 4 - 5 | 1066 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SCD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112815.1 | NP_001106286.1 | 4 - 5 | 1066 | 79 | |
| GenBank mRNA | EU272818.1 | - | 4 - 5 | 1066 | 79 | |