Assay Details
| Gene Symbol: IFT27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126366.1 | NP_001119838.1 | 5 - 6 | 352 | 70 | |
| XM_012158633.3 | XP_012014023.2 | 5 - 6 | 899 | 70 | ||
| XM_012158642.3 | XP_012014032.2 | 4 - 5 | 676 | 70 | ||
| XM_015093761.2 | XP_014949247.2 | 4 - 5 | 839 | 70 | ||
| GenBank mRNA | EU596591.1 | - | 5 - 6 | 352 | 70 | |
More Information
Set Membership: |
|

IFT27
Genomic Map