Assay Details
| Gene Symbol: SPARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166202.1 | NP_001159674.1 | 4 - 5 | 335 | 67 | |
| XM_012177565.3 | XP_012032955.1 | 5 - 6 | 67 | |||
| GenBank mRNA | GQ221054.1 | - | 4 - 5 | 335 | 67 | |
More Information
Set Membership: |
|

SPARC
Genomic Map