Assay Details
| Gene Symbol: AK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285802.2 | NP_001272731.1 | 3 - 4 | 287 | 84 | |
| GenBank mRNA | FE023570.1 | - | 2 - 3 | 222 | 84 | |
More Information
Set Membership: |
|

AK6
Genomic Map