Assay Details
| Gene Symbol: CD40LG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075101.1 | NP_001068569.1 | 1 - 2 | 152 | 91 | |
| GenBank mRNA | DQ054533.1 | - | 1 - 2 | 152 | 91 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD40LG| Gene Symbol: CD40LG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075101.1 | NP_001068569.1 | 1 - 2 | 152 | 91 | |
| GenBank mRNA | DQ054533.1 | - | 1 - 2 | 152 | 91 | |