Assay Details
| Gene Symbol: KLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093787.1 | NP_001087256.1 | 3 - 4 | 491 | 101 | |
| GenBank mRNA | EF521403.1 | - | 3 - 4 | 491 | 101 | |
More Information
Set Membership: |
|

KLK1
Genomic Map