Assay Details
| Gene Symbol: FABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114667.1 | NP_001108139.1 | 3 - 4 | 345 | 90 | |
| GenBank mRNA | EU301804.1 | - | 3 - 4 | 345 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FABP4| Gene Symbol: FABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114667.1 | NP_001108139.1 | 3 - 4 | 345 | 90 | |
| GenBank mRNA | EU301804.1 | - | 3 - 4 | 345 | 90 | |