Assay Details
| Gene Symbol: BSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143665.2 | NP_001137137.1 | 2 - 3 | 137 | 82 | |
| GenBank mRNA | FM211030.1 | - | 2 - 3 | 138 | 82 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: BSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143665.2 | NP_001137137.1 | 2 - 3 | 137 | 82 | |
| GenBank mRNA | FM211030.1 | - | 2 - 3 | 138 | 82 | |