Assay Details
| Gene Symbol: BSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143665.2 | NP_001137137.1 | 4 - 5 | 423 | 69 | |
| GenBank mRNA | FM211030.1 | - | 4 - 5 | 424 | 69 | |
More Information
Set Membership: |
|

BSP1
Genomic Map