Assay Details
| Gene Symbol: AIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162567.1 | NP_001156039.1 | 2 - 3 | 343 | 86 | |
| XM_027961104.1 | XP_027816905.1 | 2 - 3 | 360 | 86 | ||
| GenBank mRNA | FJ969406.1 | - | 2 - 3 | 343 | 86 | |
More Information
Set Membership: |
|

AIMP2
Genomic Map