Assay Details
| Gene Symbol: SQSTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163043.1 | NP_001156515.1 | 7 - 8 | 1161 | 69 | |
| GenBank mRNA | EU366490.1 | - | 1 - 2 | 202 | 69 | |
| FJ958359.1 | - | 7 - 8 | 1161 | 69 | ||
More Information
Set Membership: |
|

SQSTM1
Genomic Map