Assay Details
| Gene Symbol: MMP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166179.1 | NP_001159651.1 | 6 - 7 | 919 | 70 | |
| GenBank mRNA | AF267157.1 | - | 1 - 2 | 48 | 70 | |
| GQ221063.1 | - | 6 - 7 | 919 | 70 | ||
More Information
Set Membership: |
|

MMP13
Genomic Map