Assay Details
| Gene Symbol: H2AZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009270.1 | NP_001009270.1 | 2 - 3 | 188 | 86 | |
| GenBank mRNA | AY074805.1 | - | 2 - 3 | 188 | 86 | |
More Information
Set Membership: |
|

H2AZ1
Genomic Map