Assay Details
| Gene Symbol: IL12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009736.1 | NP_001009736.1 | 6 - 7 | 506 | 86 | |
| XM_012146332.3 | XP_012001722.2 | 5 - 6 | 616 | 86 | ||
| GenBank mRNA | AF173557.1 | - | 6 - 7 | 506 | 86 | |
More Information
Set Membership: |
|

IL12A
Genomic Map