Assay Details
| Gene Symbol: PON2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126348.1 | NP_001119820.1 | 2 - 3 | 143 | 75 | |
| GenBank mRNA | EU579457.1 | - | 2 - 3 | 143 | 75 | |
More Information
Set Membership: |
|

PON2
Genomic Map