Assay Details
| Gene Symbol: AKT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161857.1 | NP_001155329.1 | 12 - 13 | 1366 | 103 | |
| XM_027956972.1 | XP_027812773.1 | 13 - 14 | 1538 | 103 | ||
| GenBank mRNA | FJ943991.1 | - | 12 - 13 | 1366 | 103 | |
More Information
Set Membership: |
|

AKT1
Genomic Map