Assay Details
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097647.1 | NP_001091116.1 | 2 - 3 | 299 | 70 | |
| GenBank mRNA | DQ480756.1 | - | 2 - 3 | 299 | 70 | |
| GQ860955.1 | - | 2 - 3 | 299 | 70 | ||
| JX263305.1 | - | 2 - 3 | 320 | 70 | ||
More Information
Set Membership: |
|

LYZ
Genomic Map