Assay Details
| Gene Symbol: KCNK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199790.1 | NP_001186719.1 | 2 | 420 | 115 | |
| GenBank mRNA | HQ342699.1 | - | 2 | 420 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNK3| Gene Symbol: KCNK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199790.1 | NP_001186719.1 | 2 | 420 | 115 | |
| GenBank mRNA | HQ342699.1 | - | 2 | 420 | 115 | |
Set Membership: |
|