Assay Details
| Gene Symbol: BCL2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009226.2 | NP_001009226.1 | 2 | 723 | 87 | |
| XM_012188083.3 | XP_012043473.1 | 3 | 1807 | 87 | ||
| XM_027976356.1 | XP_027832157.1 | 3 | 900 | 87 | ||
| XM_027976357.1 | XP_027832158.1 | 3 | 1825 | 87 | ||
| XM_027976358.1 | XP_027832159.1 | 3 | 879 | 87 | ||
| GenBank mRNA | AF164517.1 | - | 2 | 723 | 87 | |
More Information
Set Membership: |
|

BCL2L1
Genomic Map