Assay Details
| Gene Symbol: SSTR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009265.1 | NP_001009265.1 | 2 | 518 | 95 | |
| GenBank mRNA | AJ441116.1 | - | 2 | 518 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSTR5| Gene Symbol: SSTR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009265.1 | NP_001009265.1 | 2 | 518 | 95 | |
| GenBank mRNA | AJ441116.1 | - | 2 | 518 | 95 | |
Set Membership: |
|