Assay Details
| Gene Symbol: XCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009427.1 | NP_001009427.1 | 3 | 316 | 103 | |
| GenBank mRNA | AB103064.1 | - | 3 | 316 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: XCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009427.1 | NP_001009427.1 | 3 | 316 | 103 | |
| GenBank mRNA | AB103064.1 | - | 3 | 316 | 103 | |
Set Membership: |
|