Assay Details
| Gene Symbol: TEKT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012096691.3 | XP_011952081.1 | 4 - 5 | 628 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEKT2| Gene Symbol: TEKT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012096691.3 | XP_011952081.1 | 4 - 5 | 628 | 55 | |
Set Membership: |
|