Assay Details
| Gene Symbol: CCDC134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007021.4 | XP_004007070.1 | 2 - 3 | 245 | 94 | |
| XM_012175707.3 | XP_012031097.1 | 2 - 3 | 245 | 94 | ||
More Information
Set Membership: |
|

CCDC134
Genomic Map