Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097311.2 | XP_014952797.2 | 12 - 13 | 1562 | 100 | |
| XM_027972311.1 | XP_027828112.1 | 12 - 13 | 1562 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DOP1A| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097311.2 | XP_014952797.2 | 12 - 13 | 1562 | 100 | |
| XM_027972311.1 | XP_027828112.1 | 12 - 13 | 1562 | 100 | ||