Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097311.2 | XP_014952797.2 | 15 - 16 | 2452 | 81 | |
| XM_027972311.1 | XP_027828112.1 | 15 - 16 | 2452 | 81 | ||
| XM_027972312.1 | XP_027828113.1 | 14 - 15 | 2422 | 81 | ||

DOP1A
Genomic Map