Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097311.2 | XP_014952797.2 | 27 - 28 | 6009 | 125 | |
| XM_027972311.1 | XP_027828112.1 | 27 - 28 | 6009 | 125 | ||
| XM_027972312.1 | XP_027828113.1 | 26 - 27 | 5979 | 125 | ||
More Information
Set Membership: |
|

DOP1A
Genomic Map