Assay Details
| Gene Symbol: NT5DC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011184.4 | XP_004011233.2 | 9 - 10 | 1071 | 77 | |
| XM_012182548.3 | XP_012037938.2 | 9 - 10 | 1037 | 77 | ||
More Information
Set Membership: |
|

NT5DC1
Genomic Map