Assay Details
| Gene Symbol: RPS27L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010547.3 | XP_004010596.1 | 2 - 3 | 222 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS27L| Gene Symbol: RPS27L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010547.3 | XP_004010596.1 | 2 - 3 | 222 | 78 | |
Set Membership: |
|