Assay Details
| Gene Symbol: SLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979166.1 | XP_027834967.1 | 2 - 3 | 200 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027979166.1 transcripts.
Set Membership: |
|

SLN
Genomic Map