Assay Details
| Gene Symbol: ORC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014967.3 | XP_004015016.2 | 4 - 5 | 731 | 70 | |
| XM_027977796.1 | XP_027833597.1 | 4 - 5 | 476 | 70 | ||
More Information
Set Membership: |
|

ORC6
Genomic Map