Assay Details
| Gene Symbol: PLLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015021.4 | XP_004015070.1 | 2 - 3 | 444 | 81 | |
| XM_012189822.2 | XP_012045212.1 | 2 - 3 | 589 | 81 | ||
More Information
Set Membership: |
|

PLLP
Genomic Map