Assay Details
| Gene Symbol: TEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098945.2 | XP_014954431.1 | 2 - 3 | 216 | 69 | |
| XM_015098946.2 | XP_014954432.1 | 2 - 3 | 217 | 69 | ||
| XM_015098947.2 | XP_014954433.1 | 2 - 3 | 230 | 69 | ||
| XM_027974105.1 | XP_027829906.1 | 2 - 3 | 443 | 69 | ||
More Information
Set Membership: |
|

TEN1
Genomic Map