Assay Details
| Gene Symbol: LOC101117554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020724.3 | XP_004020773.1 | 3 - 4 | 373 | 86 | |
| Gene Symbol: LOC114110580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961470.1 | XP_027817271.1 | 3 - 4 | 366 | 86 | |
More Information
Set Membership: |
|

LOC101117554
Genomic Map