Assay Details
| Gene Symbol: NSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021782.4 | XP_004021831.1 | 5 - 6 | 1603 | 58 | |
| XM_012105930.3 | XP_011961320.1 | 5 - 6 | 1603 | 58 | ||
| XM_012105933.3 | XP_011961323.1 | 5 - 6 | 1603 | 58 | ||
More Information
Set Membership: |
|

NSD3
Genomic Map