Assay Details
| Gene Symbol: IL1RAPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963506.1 | XP_027819307.1 | 9 - 10 | 1234 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL1RAPL2| Gene Symbol: IL1RAPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963506.1 | XP_027819307.1 | 9 - 10 | 1234 | 82 | |
Set Membership: |
|