Assay Details
| Gene Symbol: LOC101118866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967501.1 | XP_027823302.1 | 1 - 2 | 325 | 80 | |
| XM_027967503.1 | XP_027823304.1 | 1 - 2 | 47 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027967503.1 transcripts.
Set Membership: |
|

LOC101118866
Genomic Map