Assay Details
| Gene Symbol: LOC114112026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963709.1 | XP_027819510.1 | 2 - 3 | 166 | 73 | |
| Gene Symbol: PDE8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971477.1 | XP_027827278.1 | 6 - 7 | 555 | 73 | |
More Information
Set Membership: |
|

LOC114112026
Genomic Map