Assay Details
| Gene Symbol: KIF27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012117525.3 | XP_011972915.2 | 7 - 8 | 2155 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIF27| Gene Symbol: KIF27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012117525.3 | XP_011972915.2 | 7 - 8 | 2155 | 70 | |
Set Membership: |
|