Assay Details
| Gene Symbol: TTBK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010488.4 | XP_004010537.2 | 13 - 14 | 2213 | 63 | |
| XM_012181291.2 | XP_012036681.2 | 12 - 13 | 2172 | 63 | ||
| XM_012181295.2 | XP_012036685.2 | 12 - 13 | 1980 | 63 | ||
More Information
Set Membership: |
|

TTBK2
Genomic Map