Assay Details
| Gene Symbol: ST3GAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183518.2 | XP_012038908.1 | 3 - 4 | 1323 | 70 | |
| XM_012183520.2 | XP_012038910.1 | 2 - 3 | 1171 | 70 | ||
More Information
Set Membership: |
|

ST3GAL1
Genomic Map