Assay Details
| Gene Symbol: SIRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019744.4 | XP_004019793.1 | 5 - 6 | 811 | 64 | |
| XM_027959983.1 | XP_027815784.1 | 5 - 6 | 716 | 64 | ||
More Information
Set Membership: |
|

SIRT3
Genomic Map