Assay Details
| Gene Symbol: SCML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106304.3 | XP_011961694.1 | 5 - 6 | 619 | 101 | |
| XM_012106306.3 | XP_011961696.1 | 4 - 5 | 538 | 101 | ||
| XM_012106307.3 | XP_011961697.1 | 4 - 5 | 535 | 101 | ||
| XM_027963005.1 | XP_027818806.1 | 5 - 6 | 619 | 101 | ||
More Information
Set Membership: |
|

SCML1
Genomic Map