Assay Details
| Gene Symbol: SCML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106304.3 | XP_011961694.1 | 6 - 7 | 1173 | 96 | |
| XM_012106306.3 | XP_011961696.1 | 5 - 6 | 1092 | 96 | ||
| XM_012106307.3 | XP_011961697.1 | 5 - 6 | 1089 | 96 | ||
| XM_012106308.3 | XP_011961698.1 | 6 - 7 | 669 | 96 | ||
More Information
Set Membership: |
|

SCML1
Genomic Map