Assay Details
| Gene Symbol: LOC443423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002637.4 | XP_004002686.1 | 2 - 3 | 331 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC443423| Gene Symbol: LOC443423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002637.4 | XP_004002686.1 | 2 - 3 | 331 | 71 | |
Set Membership: |
|