Assay Details
| Gene Symbol: LOC101119130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968088.1 | XP_027823889.1 | 8 - 9 | 1263 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101119130| Gene Symbol: LOC101119130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968088.1 | XP_027823889.1 | 8 - 9 | 1263 | 55 | |
Set Membership: |
|