Assay Details
| Gene Symbol: SUV39H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099594.2 | XP_014955080.2 | 5 - 6 | 1208 | 74 | |
| XM_015099595.2 | XP_014955081.1 | 4 - 5 | 1033 | 74 | ||
More Information
Set Membership: |
|

SUV39H2
Genomic Map