Assay Details
| Gene Symbol: LOC101105780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101638.2 | XP_014957124.2 | 4 - 5 | 903 | 53 | |
| XM_027956516.1 | XP_027812317.1 | 3 - 4 | 1488 | 53 | ||
| XM_027956517.1 | XP_027812318.1 | 3 - 4 | 1490 | 53 | ||
| XM_027956518.1 | XP_027812319.1 | 4 - 5 | 830 | 53 | ||
More Information
Set Membership: |
|

LOC101105780
Genomic Map